Product Details

SNP ID
rs147131448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12624672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGAAAAATGGGAAGACCCGAA[C/T]GTTGAAGATCAACACAAAAACCAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF791 PubMed Links

Gene Details

Gene
ZNF791
Gene Name
zinc finger protein 791
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153358.2 285 Silent Mutation AAC,AAT N51N NP_699189.2
XM_017026412.1 285 Silent Mutation AAC,AAT N19N XP_016881901.1

View Full Product Details