Product Details

SNP ID
rs147175837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58368256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGGCGCACTCGTACGGCTTCTC[C/G]CCGGCTCCCGACCCCCGTCGGGGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100419840 PubMed Links

Gene Details

Gene
LOC100419840
Gene Name
zinc finger protein 446 pseudogene
There are no transcripts associated with this gene.

Gene
LOC105372483
Gene Name
uncharacterized LOC105372483
There are no transcripts associated with this gene.

Gene
ZNF497
Gene Name
zinc finger protein 497
There are no transcripts associated with this gene.

Gene
ZNF837
Gene Name
zinc finger protein 837
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138466.1 2066 Silent Mutation GGC,GGG G359G NP_612475.1
XM_017026242.1 2066 Silent Mutation GGC,GGG G359G XP_016881731.1

View Full Product Details