Product Details

SNP ID
rs147202237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38803773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTCAGCTGTTGATGGCAATGT[C/T]CGGGACCCTGAACGATGGACAGAAG
Phenotype
MIM: 602518
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS4 PubMed Links

Gene Details

Gene
LGALS4
Gene Name
galectin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006149.3 723 Missense Mutation GAA,GGA E170G NP_006140.1
XM_011526973.1 723 Intron XP_011525275.1
XM_011526974.1 723 Missense Mutation GAA,GGA E39G XP_011525276.1

View Full Product Details