Product Details

SNP ID
rs147438493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39998362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCCTCTTCACGGGCCTCCAAA[A/T]GACTTTCTCATTTTTCAAATCATTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF546 PubMed Links

Gene Details

Gene
ZNF546
Gene Name
zinc finger protein 546
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297763.1 292 Intron NP_001284692.1
NM_178544.4 292 Missense Mutation AAA,AAT K12N NP_848639.2
XM_011526898.2 292 Intron XP_011525200.1
XM_011526899.2 292 Missense Mutation AAA,AAT K12N XP_011525201.1

View Full Product Details