Product Details

SNP ID
rs147645989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18994423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCATCCTCTGTCGGAACACATC[A/G]AATGTGTCTTCTTTGTGCTCCTGCC
Phenotype
MIM: 607993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUGP2 PubMed Links

Gene Details

Gene
SUGP2
Gene Name
SURP and G-patch domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017392.4 3338 Silent Mutation TTC,TTT F1064F NP_001017392.2
NM_001321697.1 3338 Silent Mutation TTC,TTT F1064F NP_001308626.1
NM_001321698.1 3338 Silent Mutation TTC,TTT F1078F NP_001308627.1
NM_001321699.1 3338 Silent Mutation TTC,TTT F1078F NP_001308628.1
NM_014884.4 3338 Silent Mutation TTC,TTT F1064F NP_055699.2
XM_017026137.1 3338 Silent Mutation TTC,TTT F1064F XP_016881626.1

View Full Product Details