Product Details

SNP ID
rs147646924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51746274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGCGACAAAGGAGAGGACCC[A/G]TAAGGGACGACTGGACTTAATCAAG
Phenotype
MIM: 136537 MIM: 136538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FPR1 PubMed Links

Gene Details

Gene
FPR1
Gene Name
formyl peptide receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193306.1 816 Missense Mutation CGG,TGG R241W NP_001180235.1
NM_002029.3 816 Missense Mutation CGG,TGG R241W NP_002020.1
Gene
FPR2
Gene Name
formyl peptide receptor 2
There are no transcripts associated with this gene.

View Full Product Details