Product Details

SNP ID
rs147729252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45306786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGATCATGTCGTCAATGGACTG[C/G]CCTTTCTCCAACTTCTTCTCCATTT
Phenotype
MIM: 123310 MIM: 606495
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CKM PubMed Links

Gene Details

Gene
CKM
Gene Name
creatine kinase, M-type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001824.4 1285 Silent Mutation GGC,GGG G370G NP_001815.2
XM_017026240.1 1285 Silent Mutation GGC,GGG G370G XP_016881729.1
Gene
MARK4
Gene Name
microtubule affinity regulating kinase 4
There are no transcripts associated with this gene.

View Full Product Details