Product Details

SNP ID
rs147798149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57978346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTTCTCTGGTGTTGAAGTAG[C/G]GCTGAGTGACCACTAAAGGCTTTTC
Phenotype
MIM: 613905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf18 PubMed Links

Gene Details

Gene
C19orf18
Gene Name
chromosome 19 open reading frame 18
There are no transcripts associated with this gene.

Gene
ZNF606
Gene Name
zinc finger protein 606
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025027.3 2812 Silent Mutation GCC,GCG A778A NP_079303.2
XM_005259276.1 2812 Silent Mutation GCC,GCG A778A XP_005259333.1

View Full Product Details