Product Details

SNP ID
rs147866911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37228854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATATATAATCCTTCTCTAATTA[C/T]TGTATGTAAAAAAAAATTATTTTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF383 PubMed Links

Gene Details

Gene
ZNF383
Gene Name
zinc finger protein 383
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152604.1 1189 Intron NP_689817.1
XM_005258585.2 1189 Intron XP_005258642.1
XM_005258587.2 1189 Intron XP_005258644.1
XM_005258588.2 1189 Intron XP_005258645.1
XM_011526586.2 1189 Intron XP_011524888.1
XM_011526587.2 1189 Intron XP_011524889.1
XM_011526588.2 1189 UTR 5 XP_011524890.1
XM_011526589.2 1189 Intron XP_011524891.1
XM_011526590.2 1189 Intron XP_011524892.1
XM_017026422.1 1189 Intron XP_016881911.1
XM_017026423.1 1189 Intron XP_016881912.1
XM_017026424.1 1189 Intron XP_016881913.1

View Full Product Details