Product Details

SNP ID
rs148124585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21809300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGTATAATTTTTTTTATGTGTA[A/G]TAAGGCGTGAGGACCAATTAAAGAC
Phenotype
MIM: 603972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF43 PubMed Links

Gene Details

Gene
ZNF43
Gene Name
zinc finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256648.1 6565 Intron NP_001243577.1
NM_001256649.1 6565 Missense Mutation ACT,ATT T240I NP_001243578.1
NM_001256650.1 6565 Intron NP_001243579.1
NM_001256651.1 6565 Intron NP_001243580.1
NM_001256653.1 6565 Intron NP_001243582.1
NM_001256654.1 6565 Missense Mutation ACT,ATT T181I NP_001243583.1
NM_003423.3 6565 Missense Mutation ACT,ATT T246I NP_003414.2
XM_011528257.2 6565 Intron XP_011526559.1
XM_011528259.2 6565 Missense Mutation ACT,ATT T240I XP_011526561.1
XM_017027207.1 6565 Intron XP_016882696.1
XM_017027208.1 6565 Intron XP_016882697.1
XM_017027209.1 6565 Intron XP_016882698.1
XM_017027210.1 6565 Intron XP_016882699.1
XM_017027211.1 6565 Missense Mutation ACT,ATT T240I XP_016882700.1
XM_017027212.1 6565 Intron XP_016882701.1
XM_017027213.1 6565 Intron XP_016882702.1
XM_017027214.1 6565 Intron XP_016882703.1
XM_017027215.1 6565 Intron XP_016882704.1
XM_017027216.1 6565 Intron XP_016882705.1

View Full Product Details