Product Details

SNP ID
rs148218929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40717985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGGACACAACCTAGCACTGAC[A/G]GTTCCCAGACAGCACCTGGGACAGA
Phenotype
MIM: 615567 MIM: 606476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ8B PubMed Links

Gene Details

Gene
COQ8B
Gene Name
coenzyme Q8B
There are no transcripts associated with this gene.

Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025194.2 111 Missense Mutation AGT,GGT S284G NP_079470.1
XM_017027324.1 111 Silent Mutation ACA,ACG T37T XP_016882813.1

View Full Product Details