Product Details

SNP ID
rs148321762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39329021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCTTTGTGAGCTCTGCTGTCTG[C/T]ACCAGCCTGTTTTTACTCCCTGCAT
Phenotype
MIM: 604104 MIM: 612807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMFG PubMed Links

Gene Details

Gene
GMFG
Gene Name
glia maturation factor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301008.1 446 Silent Mutation GTA,GTG V71V NP_001287937.1
NM_004877.3 446 Silent Mutation GTA,GTG V112V NP_004868.1
Gene
LRFN1
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
There are no transcripts associated with this gene.

View Full Product Details