Product Details

SNP ID
rs148506413
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9185895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTGTCCATGTACCTGGTGAC[A/G]GTGCTGGGAAACCTGCTCATCATCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR7D2 PubMed Links

Gene Details

Gene
OR7D2
Gene Name
olfactory receptor family 7 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175883.2 302 Silent Mutation ACA,ACG T38T NP_787079.1

View Full Product Details