Product Details

SNP ID
rs148761753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51612261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGTCTTGATCTCCGAGTACTC[C/T]GTGGTGCTTGGGGCCTCCTGGTCCT
Phenotype
MIM: 604200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC5 PubMed Links

Gene Details

Gene
SIGLEC5
Gene Name
sialic acid binding Ig like lectin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003830.3 1765 Silent Mutation ACA,ACG T542T NP_003821.1
XM_011527438.2 1765 UTR 3 XP_011525740.1
XM_017027419.1 1765 Silent Mutation ACA,ACG T551T XP_016882908.1

View Full Product Details