Product Details

SNP ID
rs149024078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16121759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTTCATGTTTAGGGACACAGC[C/T]GGTCAGGAACGGTTTCGGACGATCA
Phenotype
MIM: 165040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB8A PubMed Links

Gene Details

Gene
RAB8A
Gene Name
RAB8A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005370.4 205 Silent Mutation GCC,GCT A65A NP_005361.2
XM_011528020.2 205 Silent Mutation GCC,GCT A65A XP_011526322.1

View Full Product Details