Product Details
- SNP ID
-
rs149291954
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:46493676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCTGGCATGGGCCCCGGCGCCC[A/G]CGCTCCCCTTCTTCTTCCTGCTTCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PNMAL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4803966] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PNMAL2
- Gene Name
- paraneoplastic Ma antigen family-like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020709.2 |
2237 |
Missense Mutation |
GCG,GTG |
A597V |
NP_065760.1 |
- Gene
- PPP5D1
- Gene Name
- PPP5 tetratricopeptide repeat domain containing 1
View Full Product Details