Product Details

SNP ID
rs149587207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39837830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCGGTGGGAGAACTCGACTGCA[C/T]AGACTAGACCATCCTAAAATAAATT
Phenotype
MIM: 604556 MIM: 134795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK1B PubMed Links

Gene Details

Gene
DYRK1B
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
There are no transcripts associated with this gene.

Gene
FBL
Gene Name
fibrillarin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001436.3 458 Missense Mutation TAT,TGT Y188C NP_001427.2
XM_005258651.2 458 Missense Mutation TAT,TGT Y187C XP_005258708.1
XM_011526623.2 458 Missense Mutation TAT,TGT Y131C XP_011524925.1
Gene
MIR6719
Gene Name
microRNA 6719
There are no transcripts associated with this gene.

View Full Product Details