Product Details

SNP ID
rs149988952
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46603327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCCAAGCTTTGGCCACAACTA[A/G]TTTCTCTTTCTGTTTGTAATCAGCA
Phenotype
MIM: 114183 MIM: 600022
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CALM3 PubMed Links

Gene Details

Gene
CALM3
Gene Name
calmodulin 3
There are no transcripts associated with this gene.

Gene
PPP5D1
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
There are no transcripts associated with this gene.

Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
There are no transcripts associated with this gene.

View Full Product Details