Product Details

SNP ID
rs150102389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11345592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGTAGATGGACCATCTTAATCA[C/G]GCCTCCTAGGTTCCCGCCTTCCTCC
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080503.2 421 Intron NP_001073972.2
XM_006722643.1 421 Intron XP_006722706.1
XM_011527678.2 421 Intron XP_011525980.2
XM_017026255.1 421 Intron XP_016881744.1
XM_017026256.1 421 Intron XP_016881745.1
XM_017026257.1 421 Intron XP_016881746.1
XM_017026258.1 421 Intron XP_016881747.1
XM_017026259.1 421 Intron XP_016881748.1
XM_017026260.1 421 Intron XP_016881749.1
XM_017026261.1 421 Intron XP_016881750.1
XM_017026262.1 421 Intron XP_016881751.1
XM_017026263.1 421 Intron XP_016881752.1
XM_017026264.1 421 Intron XP_016881753.1
Gene
PLPPR2
Gene Name
phospholipid phosphatase related 2
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145416.1 421 Missense Mutation CTG,GTG L10V NP_001138888.1
NM_001321112.1 421 Missense Mutation CTG,GTG L10V NP_001308041.1
NM_001321113.1 421 Missense Mutation CTG,GTG L10V NP_001308042.1
NM_001321114.1 421 Missense Mutation CTG,GTG L10V NP_001308043.1
NM_033408.3 421 Missense Mutation CTG,GTG L10V NP_212133.1
NM_198536.2 421 Missense Mutation CTG,GTG L10V NP_940938.1
XM_005259899.2 421 Missense Mutation CTG,GTG L10V XP_005259956.1
XM_017026769.1 421 Intron XP_016882258.1

View Full Product Details