Product Details

SNP ID
rs150238702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39706634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTGTTTCCTTGCCTGTTGGTT[A/C]GTGCGTGATAATCACAGGGACACCG
Phenotype
MIM: 607260
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LGALS14 PubMed Links

Gene Details

Gene
LGALS14
Gene Name
galectin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020129.2 276 Missense Mutation TAG,TCG *18S NP_064514.1
NM_203471.1 276 Missense Mutation TAG,TCG *47S NP_982297.1

View Full Product Details