Product Details

SNP ID
rs150305828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54866414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCTGCGGGGGACAGAACAGTG[A/C]ATAGGCAGGTAGGTCCTCCTCGGCC
Phenotype
MIM: 147045 MIM: 604947
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FCAR PubMed Links

Gene Details

Gene
FCAR
Gene Name
Fc fragment of IgA receptor
There are no transcripts associated with this gene.

Gene
KIR3DL2
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242867.1 1202 Intron NP_001229796.1
NM_006737.3 1202 Intron NP_006728.2
XM_011526940.2 1202 Missense Mutation AAT,CAT N349H XP_011525242.1
XM_017026784.1 1202 Intron XP_016882273.1

View Full Product Details