Product Details

SNP ID
rs150310136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35907261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGATACGCTGTTTCCGGGTC[A/G]CTGCTGGAGGTGAGGGGTGTTGTGG
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
There are no transcripts associated with this gene.

Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173514.1 311 Missense Mutation GCG,GTG A67V NP_001166985.1
NM_001173515.1 311 Missense Mutation GCG,GTG A66V NP_001166986.1
NM_003332.3 311 Missense Mutation GCG,GTG A78V NP_003323.1
NM_198125.2 311 Missense Mutation GCG,GTG A77V NP_937758.1

View Full Product Details