Product Details

SNP ID
rs150549756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4283070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCTAGAACTTCCCGGCAGAGG[C/G]GTGCAGCTCTATGACACCCCTTATG
Phenotype
MIM: 610481 MIM: 614715
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SHD PubMed Links

Gene Details

Gene
SHD
Gene Name
Src homology 2 domain containing transforming protein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020209.3 1883 Silent Mutation GGC,GGG G140G NP_064594.3
Gene
TMIGD2
Gene Name
transmembrane and immunoglobulin domain containing 2
There are no transcripts associated with this gene.

View Full Product Details