Product Details

SNP ID
rs150587910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35151322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTCCTGCTGGGTATCCTCATC[A/G]TCATCAGTAAGTGCGACCCATTCCT
Phenotype
MIM: 602359 MIM: 606669 MIM: 606684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXYD1 PubMed Links

Gene Details

Gene
FXYD1
Gene Name
FXYD domain containing ion transport regulator 1
There are no transcripts associated with this gene.

Gene
FXYD5
Gene Name
FXYD domain containing ion transport regulator 5
There are no transcripts associated with this gene.

Gene
FXYD7
Gene Name
FXYD domain containing ion transport regulator 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022006.1 214 Missense Mutation ATC,GTC I44V NP_071289.1

View Full Product Details