Product Details

SNP ID
rs151009341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45914515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTGCAGAAGTTGCACAGGGTC[C/G]CCAGGCCCCCGTTGGCCCCTGGCGC
Phenotype
MIM: 608228
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NANOS2 PubMed Links

Gene Details

Gene
NANOS2
Gene Name
nanos C2HC-type zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029861.2 264 Missense Mutation GCG,GGG A60G NP_001025032.1

View Full Product Details