Product Details

SNP ID
rs80358212
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:34761319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAGTACTCCACAGTGCTGAGCC[A/G]CATCTGGCTGTCTGTGGTGTTCATC
Phenotype
MIM: 605425 MIM: 604493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJB4 PubMed Links

Gene Details

Gene
GJB4
Gene Name
gap junction protein beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153212.2 723 Missense Mutation CAC,CGC H22R NP_694944.1
XM_011540679.2 723 Missense Mutation CAC,CGC H22R XP_011538981.1
Gene
GJB5
Gene Name
gap junction protein beta 5
There are no transcripts associated with this gene.

View Full Product Details