Product Details

SNP ID
rs111371881
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:943946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCGAGTTTTCTACGTGGCCAG[C/T]TTCCCCGTGGCTCTGCCACTGCAGC
Phenotype
MIM: 610770 MIM: 616765
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NOC2L PubMed Links
Additional Information
For this assay, SNP(s) [rs7523549] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015658.3 1919 Intron NP_056473.2
Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 1919 Silent Mutation AGC,AGT S613S NP_689699.2

View Full Product Details