Product Details

SNP ID
rs111613082
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:62792683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCATCAGGCTTTTACATTTAT[A/C]GCATATTTTGAGTTTAGTACAATAT
Phenotype
MIM: 611339
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG4C PubMed Links

Gene Details

Gene
ATG4C
Gene Name
autophagy related 4C cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032852.3 Intron NP_116241.2
NM_178221.2 Intron NP_835739.1
XM_005271288.2 Intron XP_005271345.1
XM_011542313.2 Intron XP_011540615.1
XM_017002600.1 Intron XP_016858089.1
XM_017002601.1 Intron XP_016858090.1

View Full Product Details