Product Details
- SNP ID
-
rs111883241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
22
- Location
-
Chr.1:97999097 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATACTATATTATTATAATTGTC[C/T]CTCTGTAATTCTAATTCTTTTGCTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR137HG
PubMed Links
Gene Details
- Gene
- MIR137HG
- Gene Name
- MIR137 host gene
There are no transcripts associated with this gene.
View Full Product Details