Product Details

SNP ID
rs112075866
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:248239123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCATGTACTTCCTCCTCAGTCA[A/G]CTGTCCCTTATGGACCTCATGCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 195 Silent Mutation CAA,CAG Q65Q NP_059974.1

View Full Product Details