Product Details
- SNP ID
-
rs112342416
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:44412124 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGACTTACACCGGGCAGCCTTCAA[A/G]ATGGAGAACTCATCCTACCTTCCCA
- Phenotype
-
MIM: 616136
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF220
PubMed Links
Gene Details
- Gene
- RNF220
- Gene Name
- ring finger protein 220
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319956.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
NP_001306885.1 |
NM_001319957.1 |
683 |
Intron |
|
|
NP_001306886.1 |
NM_018150.3 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
NP_060620.2 |
XM_005270992.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_005271049.1 |
XM_005270993.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_005271050.1 |
XM_005270996.1 |
683 |
Intron |
|
|
XP_005271053.1 |
XM_006710735.3 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_006710798.1 |
XM_011541698.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_011540000.1 |
XM_011541699.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_011540001.1 |
XM_011541700.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_011540002.1 |
XM_011541702.1 |
683 |
Intron |
|
|
XP_011540004.1 |
XM_017001623.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_016857112.1 |
XM_017001624.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_016857113.1 |
XM_017001625.1 |
683 |
Silent Mutation |
AAA,AAG |
K9K |
XP_016857114.1 |
XM_017001626.1 |
683 |
Intron |
|
|
XP_016857115.1 |
View Full Product Details