Product Details

SNP ID
rs113320975
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:85582488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTATTGTTCAAACAACTTCAT[A/G]GTCCCAGTGCTCAAAGACCTGTGGA
Phenotype
MIM: 602369 MIM: 604743
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CYR61 PubMed Links

Gene Details

Gene
CYR61
Gene Name
cysteine rich angiogenic inducer 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001554.4 931 Nonsense Mutation TAG,TGG *236W NP_001545.2
Gene
DDAH1
Gene Name
dimethylarginine dimethylaminohydrolase 1
There are no transcripts associated with this gene.

View Full Product Details