Product Details

SNP ID
rs113527964
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:86428510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGAAAAGAGGGAAAATACAT[C/T]CATTTCACACCTAATTTCCTACTGA
Phenotype
MIM: 604003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 579 Silent Mutation ATC,ATT I139I NP_006527.1
XM_011542448.2 579 Silent Mutation ATC,ATT I139I XP_011540750.1

View Full Product Details