Product Details

SNP ID
rs114413681
Assay Type
Functionally Tested
NCBI dbSNP Submissions
10
Location
Chr.1:212937689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTAGCCAAGCTCTGCAATATCT[G/T]TGGCAAATATGTTTTTTCCCCCTGG
Phenotype
MIM: 610471
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
VASH2 PubMed Links

Gene Details

Gene
VASH2
Gene Name
vasohibin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136474.2 Intron NP_001129946.1
NM_001136475.2 Intron NP_001129947.1
NM_001301056.1 Intron NP_001287985.1
NM_024749.4 Intron NP_079025.2
XM_011509987.2 Intron XP_011508289.1
XM_011509988.2 Intron XP_011508290.1
XM_011509989.2 Intron XP_011508291.1
XM_011509991.2 Intron XP_011508293.2
XM_011509994.1 Intron XP_011508296.1
XM_011509995.1 Intron XP_011508297.1
XM_011509997.1 Intron XP_011508299.1
XM_017002351.1 Intron XP_016857840.1
XM_017002352.1 Intron XP_016857841.1

View Full Product Details