Product Details

SNP ID
rs114553184
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:25832097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATAGCTGCCTTCCTCACCGCAG[A/G]TGTTTCTGCCTCTTAAGGATAGATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUNIP PubMed Links

Gene Details

Gene
AUNIP
Gene Name
aurora kinase A and ninein interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287490.1 1160 Silent Mutation CAC,CAT H369H NP_001274419.1
NM_024037.2 1160 Intron NP_076942.1
XM_017002300.1 1160 Intron XP_016857789.1
Gene
LOC646471
Gene Name
uncharacterized LOC646471
There are no transcripts associated with this gene.

Gene
MTFR1L
Gene Name
mitochondrial fission regulator 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099625.1 1160 UTR 3 NP_001093095.1
NM_001099626.1 1160 UTR 3 NP_001093096.1
NM_001099627.1 1160 UTR 3 NP_001093097.1
NM_019557.5 1160 UTR 3 NP_062457.3

View Full Product Details