Product Details

SNP ID
rs114659029
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:162852252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTATGACAACCAGAAATATCT[A/C]TGGACAGTGCTGGATGTTTTCTGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC190 PubMed Links

Gene Details

Gene
CCDC190
Gene Name
coiled-coil domain containing 190
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178550.4 6215 Intron NP_848645.3
XM_005245125.3 6215 UTR 3 XP_005245182.1
XM_006711289.3 6215 UTR 3 XP_006711352.1
XM_011509477.2 6215 UTR 3 XP_011507779.1

View Full Product Details