Product Details

SNP ID
rs114681899
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:178103517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCTGTCTCCCCACACATAATG[A/G]ATTATCACGAAATTTTGGATCAGAC
Phenotype
MIM: 606136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASAL2 PubMed Links

Gene Details

Gene
RASAL2
Gene Name
RAS protein activator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004841.3 Intron NP_004832.1
NM_170692.2 Intron NP_733793.2
XM_005245622.3 Intron XP_005245679.1
XM_011510166.2 Intron XP_011508468.1
XM_011510167.2 Intron XP_011508469.1
XM_017002849.1 Intron XP_016858338.1
XM_017002850.1 Intron XP_016858339.1
XM_017002851.1 Intron XP_016858340.1
XM_017002852.1 Intron XP_016858341.1
XM_017002853.1 Intron XP_016858342.1
XM_017002854.1 Intron XP_016858343.1
XM_017002855.1 Intron XP_016858344.1
Gene
RASAL2-AS1
Gene Name
RASAL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details