Product Details

SNP ID
rs114755789
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:42736473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCCAGACCCTTCCACCTGGCA[C/T]TCTGGGCTCTCCACACCCAGACCCT
Phenotype
MIM: 610036 MIM: 610339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN19 PubMed Links

Gene Details

Gene
CLDN19
Gene Name
claudin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123395.1 Intron NP_001116867.1
NM_001185117.1 Intron NP_001172046.1
NM_148960.2 Intron NP_683763.2
Gene
P3H1
Gene Name
prolyl 3-hydroxylase 1
There are no transcripts associated with this gene.

View Full Product Details