Product Details

SNP ID
rs114880730
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:22658455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCCACCACCTCCCACCTCCAGC[C/T]CAGTTACTCTGCCTCCCAGCATCCT
Phenotype
MIM: 120570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QB PubMed Links

Gene Details

Gene
C1QB
Gene Name
complement C1q B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000491.3 Intron NP_000482.3
XM_011542059.2 Intron XP_011540361.1

View Full Product Details