Product Details

SNP ID
rs114944537
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:42734068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTTCCCCAGAAGCCTGATACG[A/C]AGTGGCCAGGGAGACTGAGTCCCGG
Phenotype
MIM: 610036
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLDN19 PubMed Links

Gene Details

Gene
CLDN19
Gene Name
claudin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123395.1 1884 UTR 3 NP_001116867.1
NM_001185117.1 1884 UTR 3 NP_001172046.1
NM_148960.2 1884 UTR 3 NP_683763.2

View Full Product Details