Product Details

SNP ID
rs114973477
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:26162219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTTCCTTCTGGAACTCAGTTT[A/C]GCGTCTGAACCTTGGGGAGGTGGAA
Phenotype
MIM: 616698
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM110D PubMed Links

Gene Details

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024869.2 1115 UTR 3 NP_079145.2
Gene
ZNF593
Gene Name
zinc finger protein 593
There are no transcripts associated with this gene.

View Full Product Details