Product Details

SNP ID
rs115231484
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:118884261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGCACCCATTCTGGGCCTCCC[A/T]CCACAGAGTTCACCCAGTTCAGAGT
Phenotype
MIM: 604127
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBX15 PubMed Links

Gene Details

Gene
TBX15
Gene Name
T-box 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152380.2 2733 UTR 3 NP_689593.2
XM_005271161.3 2733 UTR 3 XP_005271218.1
XM_005271162.1 2733 UTR 3 XP_005271219.1
XM_011542052.2 2733 UTR 3 XP_011540354.1

View Full Product Details