Product Details

SNP ID
rs115340601
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:36798421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCATCCAAGCAAGCTTTCATG[C/T]TTCCCATCACCTCTCAGTGCCTGCG
Phenotype
MIM: 138243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIK3 PubMed Links

Gene Details

Gene
GRIK3
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000831.3 6325 UTR 3 NP_000822.2

View Full Product Details