Product Details

SNP ID
rs115359084
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:38019064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCATATTCTGTGTTCTAGAAAAT[C/T]GAAAGACTAAAATCAGAGCTCCATC
Phenotype
MIM: 609440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP11 PubMed Links

Gene Details

Gene
UTP11
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016037.3 373 Silent Mutation ATC,ATT I116I NP_057121.2
XM_011541555.2 373 Silent Mutation ATC,ATT I47I XP_011539857.1

View Full Product Details