Product Details
- SNP ID
-
hCV165916991
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145892417 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGGAAGAATCGAGGAAGATAAC[T/C]CTCATCCTTTCCAAACTGTGAGGGA
- Phenotype
-
MIM: 604042
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD35
PubMed Links
Gene Details
- Gene
- ANKRD35
- Gene Name
- ankyrin repeat domain 35
There are no transcripts associated with this gene.
- Gene
- ITGA10
- Gene Name
- integrin subunit alpha 10
There are no transcripts associated with this gene.
View Full Product Details