Product Details

SNP ID
rs115595421
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:37811345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGTCTCACAGCACAGGGCTGA[C/T]TGAGCGGAAAAACGCTGCCAGGCCA
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
There are no transcripts associated with this gene.

Gene
MANEAL
Gene Name
mannosidase endo-alpha like
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005955.2 6358 UTR 3 NP_005946.2
XM_011541491.2 6358 UTR 3 XP_011539793.1
XM_011541493.2 6358 Intron XP_011539795.1
XM_011541494.1 6358 UTR 3 XP_011539796.1
Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
There are no transcripts associated with this gene.

View Full Product Details