Product Details
- SNP ID
-
hCV165934839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145826557 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGCCCAGTGCAGCCGGGTATTGC[G/A]AATGCTTAGATATCCCCGGTACATC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
POLR3C
PubMed Links
Gene Details
- Gene
- POLR3C
- Gene Name
- RNA polymerase III subunit C
There are no transcripts associated with this gene.
- Gene
- RNF115
- Gene Name
- ring finger protein 115
There are no transcripts associated with this gene.
View Full Product Details