Product Details

SNP ID
rs116066291
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:114773507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTTTTATTTTGTGTGTGTTTT[G/T]GGCTGATTATATTTAATAAAAGCTA
Phenotype
MIM: 611656
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIKE1 PubMed Links

Gene Details

Gene
SIKE1
Gene Name
suppressor of IKBKE 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102396.1 1468 UTR 3 NP_001095866.1
NM_025073.2 1468 UTR 3 NP_079349.2

View Full Product Details