Product Details

SNP ID
rs116118596
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:197155221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACAGTAACTATAAAGGCCACAA[C/T]AGAGATTTTTAAAATGATATATAAA
Phenotype
MIM: 605481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
There are no transcripts associated with this gene.

Gene
ZBTB41
Gene Name
zinc finger and BTB domain containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194314.2 6937 UTR 3 NP_919290.2
XM_011509507.2 6937 Intron XP_011507809.1
XM_017001205.1 6937 Intron XP_016856694.1

View Full Product Details