Product Details

SNP ID
rs116287745
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:153994233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATTTCAGCAAAAGTAGCTGTT[A/G]TACCTCAAGATTTAGTAGTGGTTTC
Phenotype
MIM: 602672 MIM: 603702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159484.1 Intron NP_001152956.1
NM_207308.2 Intron NP_997191.2
XM_011510122.1 Intron XP_011508424.1
XM_011510123.1 Intron XP_011508425.1
XM_011510124.1 Intron XP_011508426.1
XM_017002788.1 Intron XP_016858277.1
XM_017002789.1 Intron XP_016858278.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RPS27
Gene Name
ribosomal protein S27
There are no transcripts associated with this gene.

View Full Product Details